Hollow fibre filter modules, PES membrane, MidiKros®
Supplier: Spectrum Laboratories
D04-P20U-10-S D04-P20U-10-N D06-P20U-10-S D06-P20U-10-N D02-P20U-10-N D02-P20U-10-S
SPECD04-P20U-10-SEA 730 EUR
SPECD04-P20U-10-S SPECD04-P20U-10-N SPECD06-P20U-10-S SPECD06-P20U-10-N SPECD02-P20U-10-N SPECD02-P20U-10-S
Hollow fibre filter modules, PES membrane, MidiKros®
Filters Tangential Flow Systems
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • The filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • TC filters have FLL/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information