Visible and UV/visible spectrophotometers, GENESYS™
Supplier: Thermo Fisher Scientific
840-297300 840-297400 840-298300 840-298400 840-300300 840-300400 840-302400 840-301300 840-301400 840-300000
634-1072EA 0 EUR
634-1072 634-1073 634-1074 634-1075 634-1078 634-1079 634-1081 634-1082 634-1083 SPET840-300000
Visible and UV/visible spectrophotometers, GENESYS™
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Genesys Visible and UV/Visible spectrophotometers provide both exceptional performance with a contemporary design.
  • Start immediately with the simple to use touch screen start menu
  • Exchange samples with ease via large and accessible sample compartment
  • Save and print your data fast any way you want it with multiple data export options
  • Genesys 140, 150, 180 and BioMate 160 are Wi-Fi compatible with optional USB adapter
The Biomate 160 includes Life Science application software package. Software includes pre-programmed life science methods such as NucleicMore Product Information