Hot air tool, HG 2120 E
Supplier: STEINEL
351403
STEI351403EA 168 EUR
STEI351403
Hot air tool, HG 2120 E
Heaters Hot Air Dryers
Electronically controlled single-hand operation tool is ideal for heat-shrinking, shaping, welding or stripping process depending on nozzles attached.
  • Ceramic heating element
  • Optimised weight balance
  • Nine steps temperature setting by thumbwheel, ranginf from 80 to 630 °C
  • Three steps airflow rate adjustment from 150 to 500 L/min