Cole-Parmer® ST-200 Series Large-Capacity Magnetic Stirrers
Supplier: Cole Parmer
04807-74
STUA04807-74EA 1620 EUR
STUA04807-74
Cole-Parmer® ST-200 Series Large-Capacity Magnetic Stirrers
Stirrers Magnetic Stirrers
Powerful magnets for strong magnetic coupling.
  • Large plate area accommodates one large vessel up to 30 L
  • Easy-to-use stirring control dial
  • Robust, durable construction with stainless steel top
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
These general-purpose stirrers are designed with safety as well as performance in mind. Powerful magnets and motor give stirring speeds up to 600 min⁻¹ and can mix up to 30 L easily. The electronic feedback speed control accurately maintains the set speed.
Units feature BioCoteMore Product Information