Cole-Parmer® RV-400 Rotary Evaporators
Supplier: Cole Parmer
52410-00 52411-97 52411-98 52411-99
STUA52410-00EA 17410 EUR
STUA52410-00 STUA52411-97 STUA52411-98 STUA52411-99
Cole-Parmer® RV-400 Rotary Evaporators
Evaporators Rotary Evaporators
Motorised lift provides smooth, stable lifting and lowering of the bath.
  • Set and view rotation speed, temperature and timer on the large LCD
  • PTFE sealing system offers excellent thermostability
  • PID temperature controller provides stable temperature control
  • High-efficiency drain port and injection valves
  • Small footprint takes up minimal laboratory space
These rotary evaporators with motorised lift system move the bath up or down smoothly with a simple push of a button. The control panel features up and down arrows to control the bath height, push-More Product Information