Polished Stainless Steel Mirrors
Supplier: TEKNOMEK
CRMI-PM60X45 CRMI-PM60X40 CRMI-PM18X60
TEKMCRMI-PM60X45EA 0 EUR
TEKMCRMI-PM60X45 TEKMCRMI-PM60X40 TEKMCRMI-PM18X60
Polished Stainless Steel Mirrors
Mirrors
Polished stainless steel mirrors are hygienic mirrors made from 304-grade stainless steel, built to handle the toughest of environments without breaking or shattering.
  • Shatterproof and safe
  • De-burred edges and rounded corners to prevent injury
  • 1,2 mm thick stainless steel
  • Choose with or without pre-drilled mounting holes
  • Vandal resistant
They afford maximum security and hygiene, ideal for your safe and conscious workplace. These steel mirrors are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durableMore Product Information