Mobile Tool Cabinets
Supplier: TEKNOMEK
SHCCCATO4D1050 SHCCCATO111050
TEKMSHCCCATO4D1050EA 0 EUR
TEKMSHCCCATO4D1050 TEKMSHCCCATO111050
Mobile Tool Cabinets
Furniture Storage Cabinets
This heavy duty mobile stainless steel tool cabinet with 3, 4 or 11 drawers has a brushed dull polish finish is excellent for tool storage within the workplace. With a floor clearance of 100 mm, there is plenty of room for a mop or broom for easy cleaning below the tool cabinet.
  • Easy tool organisation
  • Fully welded 304L-grade stainless steel
  • Brushed dull polish finish
  • Stainless steel castors (2 fixed and 2 locking swivel)
  • Lockable drawers
Designed with lockable drawers of varying sizes this storage cabinet provides easy and safe compartmentalisationMore Product Information