Storage tray rack, for upright freezers
Supplier: TENAK
TE29428 TE29432 TE29100 TE29104 TE29108 TE29120 TE29124 TE29128 TE29132 TE29136 TE29150 TE29154 TE29158 TE29162 TE29200 TE29204 TE29208 TE29220 TE29224 TE29228 TE29232 TE29236 TE29250 TE29254 TE29258 TE29262 TE29300 TE29304 TE29308 TE29320 TE29324 TE29328 TE29332 TE29336 TE29350 TE29354 TE29358 TE29362 TE29400 TE29404 TE29408 TE29420 TE29424 TE29436 TE29450 TE29454 TE29458 TE29462
TENATE29428EA 249 EUR
TENATE29428 TENATE29432 TENATE29100 TENATE29104 TENATE29108 TENATE29120 TENATE29124 TENATE29128 TENATE29132 TENATE29136 TENATE29150 TENATE29154 TENATE29158 TENATE29162 TENATE29200 TENATE29204 TENATE29208 TENATE29220 TENATE29224 TENATE29228 TENATE29232 TENATE29236 TENATE29250 TENATE29254 TENATE29258 TENATE29262 TENATE29300 TENATE29304 TENATE29308 TENATE29320 TENATE29324 TENATE29328 TENATE29332 TENATE29336 TENATE29350 479-1323 TENATE29358 TENATE29362 TENATE29400 TENATE29404 TENATE29408 TENATE29420 TENATE29424 TENATE29436 TENATE29450 TENATE29454 TENATE29458 TENATE29462
Storage tray rack, for upright freezers
Racks Cryogenic Box Racks
These stainless steel single and double tray rack have one and two drawers respectively.
  • Ideal for odd sized samples
  • With lid and a handle on the front