Standard rack for upright freezers for (HxD) 5x7=35 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, dimensions (HxDxW) 181x631x135mm 1 * 1 items
Supplier: TENAK
Standard rack for upright freezers for (HxD) 5x7=35 TENAK Microscope slide boxes (87x128mm), 32mm high, stainless steel, dimensions (HxDxW) 181x631x135mm 1 * 1 items
New Product