SURFLO® Winged Infusion Sets
Supplier: TERUMO
8SV27ELF
TERU8SV27ELFEA 94.6 EUR
TERU8SV27ELF
SURFLO® Winged Infusion Sets
Catheters
SURFLO winged infusion sets feature a precisely honed needle with a thin wall design, ensuring easy penetration and continued access with a single insertion.
The soft, flexible interlocking wings readily conform to the body of the patient, while the angle of the wings helps position the needle and reduce the risk of vessel damage during venipuncture.