Mini infrared thermometers, testo 805
Supplier: Testo
0560.8051 0563.8051
TEST0560.8051EA 53 EUR
TEST0560.8051 TEST0563.8051
Mini infrared thermometers, testo 805
Thermometers Digital Thermometers
The testo 805 mini infrared thermometer is perfect for quick surface temperature checks, e.g. for food controls in supermarkets, for recording the temperature of incoming goods, or for scanning entire refrigerated sections.
  • Scan mode for long-term measurement
  • Pocket size measuring instrument: Compact for easy handling
  • 1:1 optics: Suitable for close-up temperature measurements
TopSafe protective case (only included for article TEST0563.8051) protects from water and dirt in accordance with IP 65. This protective case is of great benefit, e.g. in theMore Product Information