Sterile cotton swab with polystyrene handle, sterile, dry collection and transport system, 1swab/tube, 100 tubes/box, 5 boxes/case 1 * 500 items
Supplier: Texwipe
Sterile cotton swab with polystyrene handle, sterile, dry collection and transport system, 1swab/tube, 100 tubes/box, 5 boxes/case 1 * 500 items