Rotors for Medium and Large Capacity Concentrators, Savant™ Speedvac™
Supplier: THERMO SAVANT
RH64-11 RH6-50 MPTR8-210 RH10-15 RH192-15 RH200-12 RH40-11 RH48-18-125 RH52-15 RHDW2MP RHSW6MP PRO-50 PRO-20 PRO-10S RH12-28 RH100-8 RH100-6 RH50-28-60 RH88-16-125 RH8-50 RH8-200 RH8-17.5 RH72-12 RH6-25 RH6-18-150 RH24-15 RH60-12-40 RH24-18 RH48-50U RH4-18-150 RH4-100 RH32-18-150
THSARH64-11EA 1160 EUR
THSARH64-11 THSARH6-50 THSAMPTR8-210 THSARH10-15 THSARH192-15 THSARH200-12 THSARH40-11 THSARH48-18-125 THSARH52-15 THSARHDW2MP THSARHSW6MP THSAPRO-50 THSAPRO-20 THSAPRO-10S THSARH12-28 THSARH100-8 THSARH100-6 THSARH50-28-60 THSARH88-16-125 THSARH8-50 THSARH8-200 THSARH8-17.5 THSARH72-12 THSARH6-25 THSARH6-18-150 THSARH24-15 THSARH60-12-40 THSARH24-18 THSARH48-50U THSARH4-18-150 THSARH4-100 THSARH32-18-150
Rotors for Medium and Large Capacity Concentrators, Savant™ Speedvac™
Centrifuges Rotors
These rotors can be combined with medium and large capacity Savant™ SpeedVac™ concentrators.
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into the concentrator