Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Supplier: TOURNAIRE-EMBALLAGE
6951S2 5076S1 4934S1 5374S1 6952S2 7133S1 7136S1 4931S1 5077S1 5240S1 5243S1 6461S1
215-2949EA 4950 EUR
215-2949 TOUR5076S1 215-2945 215-3259 TOUR6952S2 TOUR7133S1 TOUR7136S1 TOUR4931S1 TOUR5077S1 TOUR5240S1 TOUR5243S1 TOUR6461S1
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Bottles Metal Bottles
Pure aluminium.
  • Unbreakable, odorless
  • Very good temperature resistance
  • Leakproof with tamper-evident screw caps (must be odered separately)
  • UN-approved
Suitable for storage and transport of highly aggessive liquids.