Boat-combustion Quartz glass in application temperatures of up to 1050C (maximum 1600C), without handle, translucent, high chemic al resistance and purity. length 103mm, width 20mm, height 12mm. The durability or operating cycles of our Boat-combus 1 * 1 items
Supplier: TECHNICAL QUARTZ SOLUTIONS
Boat-combustion Quartz glass in application temperatures of up to 1050C (maximum 1600C), without handle, translucent, high chemic al resistance and purity. length 103mm, width 20mm, height 12mm. The durability or operating cycles of our Boat-combus 1 * 1 items