Lid for quartz inceneration crucible in application temperatures of up to 1050C, transparent, high chemical resistance and purity. The durability or operating cycles of our quartz glass product mainly depends on applications. For application temper 1 * 1 items
Supplier: TECHNICAL QUARTZ SOLUTIONS
Lid for quartz inceneration crucible in application temperatures of up to 1050C, transparent, high chemical resistance and purity. The durability or operating cycles of our quartz glass product mainly depends on applications. For application temper 1 * 1 items