Micro burner with needle valve, cartridge type
Supplier: USBECK Laborgeräte
1423
USBE1423EA 42.8 EUR
USBE1423
Micro burner with needle valve, cartridge type
Heaters Gas Burners
Micro burner with piezo electric ignition and air regulation. Maximum temperature of 1640 °C.