Anti-IFRD2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Supplier: US Biological
036894-APC
USBI036894-APCEA 884 EUR
USBI036894-APC
Anti-IFRD2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: IFRD2 (interferon-related developmental regulator 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR