Anti-TAF10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
042574-HRP
USBI042574-HRPEA 884 EUR
USBI042574-HRP
Anti-TAF10 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: TAF10 (TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 30kDa)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR