Anti-TUBGCP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
043464-AP
USBI043464-APEA 884 EUR
USBI043464-AP
Anti-TUBGCP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TUBGCP3 (tubulin, gamma complex associated protein 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR