Anti-CCDC148 Mouse Polyclonal Antibody

Supplier: US Biological
124425
USBI124425EA 655 EUR
USBI124425
Anti-CCDC148 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCDC148 (coiled-coil domain containing 148)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CCDC148
Antigen name coiled-coil domain containing 148
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Mouse
Isotype IgG
Western blot Yes
Sequence MCAASASPDNLVFHMKNEMRNIKYKPVDYQQLRALTEAKKLASASAKLKIRKAMLTSKLSKEQTLIKQHKQVWWQEYQRLNEVRCKMESEIKSLLNEENIGNECLCDLTNFEQELSEQQCTYLKNVINPIQQLRADLKYRQHHTLQHSHPHIEFNSVKVLEEVDFVKKQLKTVFERLRLEQQRIENDLSDWSIKILDHSLEEKTNPLSELPIELESLECPYPDLKSSILSEFYKFTQKYQKKLQDFNLQLEDIYR
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR