Anti-CXCL1 Mouse Monoclonal Antibody [clone: 7A11]

Supplier: US Biological
125496
USBI125496EA 655 EUR
USBI125496
Anti-CXCL1 Mouse Monoclonal Antibody [clone: 7A11]
Antibodies
Type: Primary
Antigen: CXCL1 (chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha))
Clonality: monoclonal
Clone: 7A11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR