Anti-SPOCK2 Goat Polyclonal Antibody (Biotin)

Supplier: US Biological
145613
USBI145613EA 943 EUR
USBI145613
Anti-SPOCK2 Goat Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: SPOCK2 (sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR