Anti-AIRE Goat Polyclonal Antibody

Supplier: US Biological
A1059-88G
USBIA1059-88GEA 843 EUR
USBIA1059-88G
Anti-AIRE Goat Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot:1 ?g/ml

Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR