Anti-APEX1 Rabbit Polyclonal Antibody

Supplier: US Biological
A2298-74K
USBIA2298-74KEA 647 EUR
USBIA2298-74K
Anti-APEX1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:300-1:1000; Immunochemistry: 1:100-1:200

Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol APEX1
Environmentally Preferable
Antigen name APEX nuclease (multifunctional DNA repair enzyme) 1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification purified by immunoaffinity chromatography
Concentration ~0,1 mg/ml
Storage buffer PBS
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR