Anti-Blood Group H Antigen Mouse Monoclonal Antibody [clone: A70-A/A9]

Supplier: US Biological
B2125-22B
USBIB2125-22BEA 541 EUR
USBIB2125-22B
Anti-Blood Group H Antigen Mouse Monoclonal Antibody [clone: A70-A/A9]
Antibodies
Type: Primary
Antigen: Blood Group H Antigen
Clonality: monoclonal
Clone: A70-A/A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR