Anti-BRSK1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Supplier: US Biological
B2673-75E-PE
USBIB2673-75E-PEEA 884 EUR
USBIB2673-75E-PE
Anti-BRSK1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: BRSK1 (BR serine/threonine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR