Anti-PECAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H107]

Supplier: US Biological
C2383-01D
USBIC2383-01DEA 708 EUR
USBIC2383-01D
Anti-PECAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 7H107]
Antibodies
Do not freeze!

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 7H107
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR