Anti-Escherichia coli, J5 LPS Mouse Monoclonal Antibody [clone: 10C141]

Supplier: US Biological
E3500-28J
USBIE3500-28JEA 546 EUR
USBIE3500-28J
Anti-Escherichia coli, J5 LPS Mouse Monoclonal Antibody [clone: 10C141]
Antibodies
Type: Primary
Antigen: Escherichia coli, J5 LPS
Clonality: monoclonal
Clone: 10C141
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR