Anti-KDM2B Rabbit Polyclonal Antibody

Supplier: US Biological
K0138-40
USBIK0138-40EA 486 EUR
USBIK0138-40
Anti-KDM2B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: KDM2B (lysine (K)-specific demethylase 2B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol KDM2B
Antigen name lysine (K)-specific demethylase 2B
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
ELISA Yes
Epitope N-terminal
Form liquid
Purification purified by ammonium sulfate precipitation
Storage buffer PBS, 0,09% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR