Anti-Neuroepithelial Cell Transforming Gene 1 Goat Polyclonal Antibody

Supplier: US Biological
N2146-70E
USBIN2146-70EEA 640 EUR
USBIN2146-70E
Anti-Neuroepithelial Cell Transforming Gene 1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1: 32000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: Neuroepithelial Cell Transforming Gene 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR