Anti-N-myc Downstream Regulated Gene 1 Protein Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
N2915-52N-AP
USBIN2915-52N-APEA 884 EUR
USBIN2915-52N-AP
Anti-N-myc Downstream Regulated Gene 1 Protein Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: N-myc Downstream Regulated Gene 1 Protein
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR