Anti-OTUB1 Goat Polyclonal Antibody

Supplier: US Biological
O8490-01B
USBIO8490-01BEA 640 EUR
USBIO8490-01B
Anti-OTUB1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:64000; Western Blot: 0. 03-0.1 ?g/ml

Type: Primary
Antigen: OTUB1 (OTU deubiquitinase, ubiquitin aldehyde binding 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR