Anti-Potassium Large Conductance Calcium-activated Channel, Subfamily M, Beta Member 3 Rabbit Polyclonal Antibody

Supplier: US Biological
P5000-70D
USBIP5000-70DEA 632 EUR
USBIP5000-70D
Anti-Potassium Large Conductance Calcium-activated Channel, Subfamily M, Beta Member 3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: Potassium Large Conductance Calcium-activated Channel, Subfamily M, Beta Member 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR