Anti-Retinoic Acid Receptor Responder 1 Goat Polyclonal Antibody

Supplier: US Biological
R1691-98
USBIR1691-98EA 870 EUR
USBIR1691-98
Anti-Retinoic Acid Receptor Responder 1 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 1.0 ?g/ml

Type: Primary
Antigen: Retinoic Acid Receptor Responder 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR