Anti-MAPT Rabbit Polyclonal Antibody

Supplier: US Biological
T1030-06U
USBIT1030-06UEA 764 EUR
USBIT1030-06U
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR