Anti-TANK Rabbit Polyclonal Antibody

Supplier: US Biological
T8171-75E
USBIT8171-75EEA 449 EUR
USBIT8171-75E
Anti-TANK Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 0.5-4?g/ml; Immunoprecipitation: 10-20 ?g/ml; Immunochemistry: 10-20 ?g/ml

Type: Primary
Antigen: TANK (TRAF family member-associated NFKB activator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR