Anti-TNF Mouse Monoclonal Antibody [clone: 6A648]

Supplier: US Biological
T9160-04J
USBIT9160-04JEA 614 EUR
USBIT9160-04J
Anti-TNF Mouse Monoclonal Antibody [clone: 6A648]
Antibodies
Recommended dilution: Immunohistology: 1:10; Flow Cytometry: 1:10; Western Blot: 1:10

Type: Primary
Antigen: TNF (tumor necrosis factor)
Clonality: monoclonal
Clone: 6A648
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol TNF
Antigen name tumor necrosis factor
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 6A648
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
Flow cytometry Yes
Form liquid
Purification purified by Protein G chromatography
Concentration ~0,1 mg/ml
Storage buffer PBS, containing 0,02% sodium azide and 0,1%BSA.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR