Anti-Tumor necrosis factor receptor homolog 3 Rat Monoclonal Antibody [clone: 9H171]

Supplier: US Biological
T9161-75
USBIT9161-75EA 788 EUR
USBIT9161-75
Anti-Tumor necrosis factor receptor homolog 3 Rat Monoclonal Antibody [clone: 9H171]
Antibodies
Recommended dilution: Flow Cytometry: 25 ?g/ml

Type: Primary
Antigen: Tumor necrosis factor receptor homolog 3
Clonality: monoclonal
Clone: 9H171
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR