Anti-Uridine-5’-Diphosphoglucose Dehydrogenase Rabbit Polyclonal Antibody

Supplier: US Biological
U2279-03A
USBIU2279-03AEA 818 EUR
USBIU2279-03A
Anti-Uridine-5’-Diphosphoglucose Dehydrogenase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Uridine-5’-Diphosphoglucose Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR