Refill earplugs for "one 2 click" dispenser
Supplier: UVEX
2112.003 2112.023 2112.118 2112.022
UVEX2112.003EA 52.3 EUR
UVEX2112.003 UVEX2112.023 UVEX2112.118 UVEX2112.022
Refill earplugs for "one 2 click" dispenser
Earplugs Disposable Earplugs
Refill packs and boxes for the uvex x-fit earplug dispenser "one 2 click".
uvex x-fit: The ergonomically shaped uvex x-fit disposable earplug is characterised by very high sound absorption levels, making it suitable for use in very loud environments. The uvex x-fit foam earplug has a sealed surface, preventing foreign bodies from penetrating. The soft foam offers real comfort, even when worn for extended periods. Ideal for use in very loud environments. SNR = 37 dB.
uvex com4-fit: With its compact, ergonomically shaped design, the uvex com4-fit is comfortable even for wearers withMore Product Information