Strong anion exchange HPLC columns, PL-SAX
Supplier: VARIAN
PL1951-3802 PL1151-1802 PL1151-1803 PL1151-3802 PL1151-3803 PL1951-1502 PL1951-1503 PL1951-1802 PL1951-1803 PL1951-3803 PL1551-1502 PL1551-1503 PL1551-1802 PL1551-1803 PL1551-3102 PL1551-3103 PL1551-3702 PL1551-3703 PL1551-3802 PL1551-3803 PL1551-5102 PL1551-5103 PL1551-5702 PL1551-5703
VARIPL1951-3802EA 1060 EUR
VARIPL1951-3802 VARIPL1151-1802 VARIPL1151-1803 VARIPL1151-3802 VARIPL1151-3803 VARIPL1951-1502 VARIPL1951-1503 VARIPL1951-1802 VARIPL1951-1803 VARIPL1951-3803 VARIPL1551-1502 VARIPL1551-1503 VARIPL1551-1802 VARIPL1551-1803 VARIPL1551-3102 VARIPL1551-3103 VARIPL1551-3702 VARIPL1551-3703 VARIPL1551-3802 VARIPL1551-3803 VARIPL1551-5102 VARIPL1551-5103 VARIPL1551-5702 VARIPL1551-5703
Strong anion exchange HPLC columns, PL-SAX
Chromatography Columns
These columns are ideal for the anion exchange HPLC separations of proteins and deprotected synthetic oligonucleotides under denaturing conditions.
  • The strong anion exchange functionality and covalently linked to a chemically stable polymer that extends the operating pH range
  • Wide range of particle sizes for flexible analysis to scale-up purification
  • Small particles deliver excellent chromatographic performance
  • The anion exchange capacity is independent of pH
  • Exceptional stability for long column lifetime
For synthetic oligonucleotides, separationsMore Product Information