Stereo microscopes, ergonomic, Mantis
Supplier: VISION ENGINEERING
MCO-002
VISIMCO-002EA 280 EUR
VISIMCO-002
Stereo microscopes, ergonomic, Mantis
Microscopes
Superior imaging for a wide range of inspection and rework tasks.
  • Patented optical technology for fatigue-free viewing and superb image quality
  • Wide range of magnification options to 20x
  • Long working distances; large depths of field
  • Shadow-free true colour LED illumination
  • Choice of table stand or articulated arm
Mantis Compact excels in the low magnification range for inspection or manipulation tasks where bench magnifiers have traditionally been used. Mantis Compact has a small footprint and low nvestment cost giving a superb price/performance ratio.More Product Information