Upright Ultra-Low Temperature Freezer SU780XLE Capacity: 780 Liters (824 Quarts / 27.5 cu.ft.) Temperature Range: -86C to -20C 100% Natural Refrigerants Includes: 2 adjustable shelves 2 Year Comprehensive Warranty 7-year Stirling Cooling Technology 1 * 1 items
Supplier: VWR USA
Upright Ultra-Low Temperature Freezer SU780XLE Capacity: 780 Liters (824 Quarts / 27.5 cu.ft.) Temperature Range: -86C to -20C 100% Natural Refrigerants Includes: 2 adjustable shelves 2 Year Comprehensive Warranty 7-year Stirling Cooling Technology 1 * 1 items