Acetic acid

Supplier: VWR Collection
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

VWRS5257.9190EA 0 EUR
VWRS5257.9190
Acetic acid
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR