VWR®, Hydrometers
Supplier: VWR Collection
35125-682 35125-707 35125-729 35123-304 35123-326 35123-348 35123-370 35123-382 35123-406 35123-428 35123-450 35123-462 35125-649 35125-671 35125-751 35125-762 35125-784 35125-809 35125-831
VWRU35125-682EA 0 EUR
VWRU35125-682 VWRU35125-707 VWRU35125-729 VWRU35123-304 VWRU35123-326 VWRU35123-348 VWRU35123-370 VWRU35123-382 VWRU35123-406 VWRU35123-428 VWRU35123-450 VWRU35123-462 VWRU35125-649 VWRU35125-671 VWRU35125-751 VWRU35125-762 VWRU35125-784 VWRU35125-809 VWRU35125-831
VWR®, Hydrometers
Hydrometers
These hydrometers are designed for industries such as food and beverage, chemicals, pharmacology, transportation and refrigeration.
  • Specific gravity of petroleum products, heavy and light liquids
  • Lead free metal ballast
  • Corrections to one tenth of the smallest division
Certifications: Traceable to NIST and DAkkS. Calibration conforms with A2LA accreditions. ASTM calibrated.
Delivery information: Supplied with calibration report.