VWR®, Dialamatic Microdispensers
Supplier: VWR Collection
53506-100 53506-121
VWRU53506-100EA 0 EUR
VWRU53506-100 VWRU53506-121
VWR®, Dialamatic Microdispensers
Pipettes Repetitive Pipettes
This adjustable dispenser features a digital readout.
  • Whole or fractional microliter volumes can be set
  • Unique dual calibration system provides an accuracy and reproducibility of better than ±1%
  • Calibration mark on each bore allows a visual check for complete delivery
  • Easy to set and easy to use
  • Positive displacement pipetting of any fluid
Positive displacement piston ensures complete recovery of sample. A cutout in the barrel of the unit permits visual check for proper insertion of capillary bore. Color-coded mark on the bore lines up with stainlessMore Product Information