Bottles, serum, WHEATON®
Supplier: DWK Life Sciences
224007 224031 224033 224036 224037
216-3019EA 560 EUR
216-3019 WHEA224031 WHEA224033 WHEA224036 WHEA224037
Bottles, serum, WHEATON®
Vials
These bottles are manufactured from 400 borosilicate glass that conforms to USP Type I requirements.
  • Accepts standard stoppers and aluminum seals
  • PP bottle is used extensively for diluent solutions
  • HDPE bottle is widely used for animal health products and storage of non-medical products
  • Autoclavable at 121 °C for 20 minutes