WHEATON® COMPLETEPACK Vials with Stoppers and Seals, Amber Glass
Supplier: DWK Life Sciences
W991000-CPA W991001-CPA W991002-CPA W991003-CPA W991004-CPA W991005-CPA W991006-CPA W991007-CPA W991008-CPA W991009-CPA W991010-CPA W991011-CPA W991012-CPA W991013-CPA W991014-CPA W991015-CPA W991016-CPA W991017-CPA
WHEAW991000-CPAEA 4170 EUR
WHEAW991000-CPA WHEAW991001-CPA WHEAW991002-CPA WHEAW991003-CPA WHEAW991004-CPA WHEAW991005-CPA WHEAW991006-CPA WHEAW991007-CPA WHEAW991008-CPA WHEAW991009-CPA WHEAW991010-CPA WHEAW991011-CPA WHEAW991012-CPA WHEAW991013-CPA WHEAW991014-CPA WHEAW991015-CPA WHEAW991016-CPA WHEAW991017-CPA
WHEATON® COMPLETEPACK Vials with Stoppers and Seals, Amber Glass
Vials
Complete range of amber crimp top vials, rubber stoppers and seals that are sterilised and ready-to-use.
  • Complete manufacturing and certificate of analysis traceability
  • Container Closure Integrity (CCI) validation
  • Amber glass vials especially for photosensitive contents
Ready-to-Use, Just-in-Time, minimum 180 day shelf life from time of shipment.
Certifications: USP <788>/EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP <85>/EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻⁶ (More Product Information