High Temperature Dewar for Bruker ER 4114HT-1000 cavity. The three wall finger system made from synthetic quartz yields low thermal expansion coefficient, which results in only a small resonance frequency drift due to wall heating. This Dewar also pr 1 * 1 items
Supplier: SP Industries
High Temperature Dewar for Bruker ER 4114HT-1000 cavity. The three wall finger system made from synthetic quartz yields low thermal expansion coefficient, which results in only a small resonance frequency drift due to wall heating. This Dewar also pr 1 * 1 items